Learn More
Medchemexpress LLC Bis-NH2-PEG2 | 929-59-9 | 99.8% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY128833
Encompass_Preferred
Bis-NH2-PEG2 (PROTAC Linker 19) is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates the degradation of target proteins via the ubiquitin-proteasome system
Catalog No. 50-003-41465