Learn More
Medchemexpress LLC Bis-NH2-PEG2 (PROTAC Linker 19) | 929-59-9 | 99.8% | 100 G
Supplier: Medchemexpress LLC HY128833100G
Bis-NH2-PEG2 is a PEG-based linker utilized in the creation of protein degraders. These degraders are designed to harness the cell's natural waste disposal system, the ubiquitin-proteasome pathway, to selectively eliminate specific proteins. This product acts as a crucial connector in the construction of such protein-degrading molecules, linking a ligand for an E3 ubiquitin ligase with a ligand for a target protein.
- Serves as a foundational component for synthesizing protein degraders.
- Instrumental in promoting targeted protein degradation.
- Has a purity of 99.8%.
- Supplied in a liquid form.