Learn More
Strem, An Ascensus Company CAS# 36620-11-8. 1g. Bis(norbornadiene)rhodium(I) tetrafluoroborate, min. 96%. MFCD00671775
SDP
Supplier:  Strem, An Ascensus Company 4502301G
Encompass
CAS# 36620-11-8. 1g. Bis(norbornadiene)rhodium(I) tetrafluoroborate, min. 96%. MFCD00671775. Molecular Weight: 373.99. Molecular Formula: Rh(C7H8)2+BF4ae. Color/form: red solid Strem# 45-0230. http://www.strem.com/catalog/v/45-0230/
Catalog No. 50-901-12138