Learn More
Medchemexpress LLC Bis-PEG12-acid | 2667583-09-5 | 99.9% | C28H54O16 | 250 MG

Supplier: Medchemexpress LLC 250MGHY141020
Bis-PEG12-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is intended for research use only.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- High purity (99.9%)
- Appears as an off-white to yellow solid
- Store at 4°C, sealed, and away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.