Learn More
Medchemexpress LLC Bis-PEG2-acid | 19364-66-0 | 98.0% | 206.19 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1125595G
Encompass_Preferred
Bis-PEG2-acid is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-004-36351