Learn More
Medchemexpress LLC Bis-PEG3-NHS ester | 1314378-16-9 | 97.0% | 444.39 | C18H24N2O11 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1300871G
Encompass_Preferred
Bis-PEG3-NHS ester is a homobifunctional, noncleavable 3-unit polyethylene glycol (PEG) linker bearing two N-hydroxysuccinimide (NHS) ester groups for amine-reactive bioconjugation. It is supplied for research use and is commonly used in antibody-drug conjugate synthesis and other protein labeling applications.
  • Noncleavable 3-unit PEG spacer
  • Two amine-reactive NHS ester termini
  • Improves aqueous solubility through a hydrophilic PEG spacer
  • High purity (97.0%) suitable for research applications
  • Available in mg to gram pack sizes for lab-scale use
Catalog No. 50-002-01996