Learn More
Medchemexpress LLC Bis-PEG30-NHS ester | 95.0% | 1633.81 g·mol⁻¹ | C72H132N2O38 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16472410MG
Encompass_Preferred
Bis-PEG30-NHS ester is a homobifunctional, non-cleavable polyethylene glycol (PEG) linker containing 30 ethylene glycol units and two N-hydroxysuccinimide (NHS) ester reactive groups. It is used to conjugate antibodies and other amine-containing biomolecules in antibody-drug conjugate (ADC) synthesis and general bioconjugation. Supplied as a white to off-white powder, it has a molecular weight of 1633.81 g·mol⁻¹ and a reported purity of 95.0%.
  • Homobifunctional NHS ester termini for efficient amine coupling.
  • 30-unit PEG spacer provides hydrophilicity and distance between conjugated moieties.
  • Suitable for antibody-drug conjugate (ADC) synthesis and general bioconjugation applications.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Characterized by 1H NMR and mass spectrometry to confirm structure and purity.
Catalog No. 50-002-89293