Learn More
Medchemexpress LLC Bis-PEG4-PFP ester | 1314378-12-5 | 97.0% | 1 G

Supplier: Medchemexpress LLC HY1269981G
Bis-PEG4-PFP ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs themselves consist of two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other targeting a protein of interest. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This compound has a molecular weight of 626.39 and a chemical formula of C24H20F10O8.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Molecular weight of 626.39
- Chemical formula C24H20F10O8
- Appears as a colorless to light yellow liquid
- Density of 1.473 g/cm³
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.