Learn More
Medchemexpress LLC Bis-PEG4-PFP ester | 1314378-12-5 | 97.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126998100MG
Encompass_Preferred
Bis-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker.
  • Can be used in the synthesis of protacs.
Catalog No. 50-003-42826