Learn More
Medchemexpress LLC Bis-PEG4-PFP ester | 1314378-12-5 | 97.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY126998
Encompass_Preferred
Bis-PEG4-PFP ester is a PROTAC linker that is PEG-based and can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Consists of two distinct ligands connected by a linker
  • Targets an E3 ubiquitin ligase with one ligand
  • Targets a specific protein with the other ligand
  • Leverages the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-41468