Learn More
Medchemexpress LLC Bis-PEG5-acid 100mg | 439114-13-3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY116006100MG
Encompass_Preferred
Bis-PEG5-acid is a polyethylene glycol (PEG) based bifunctional linker with two terminal carboxylic acids. It serves as a hydrophilic spacer for PROTAC synthesis and bioconjugation, enabling formation of amide or ester bonds while improving solubility and flexibility.
  • Hydrophilic polyethylene glycol spacer.
  • Two terminal carboxylic acids for coupling reactions.
  • High purity suitable for research applications.
  • Solid form, convenient for storage and handling.
  • Available in laboratory pack sizes including 100 mg.
Catalog No. 50-002-29329