Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG6-acid | 119189-70-7 | ≥98.0% | 250 MG
SDP

Supplier:  Medchemexpress LLC HY120587250MG

Encompass_Preferred

Bis-PEG6-acid is a bifunctional polyethylene glycol (PEG) linker with two terminal carboxylic acid groups (PEG6 spacer). It is supplied as a solid, high-purity reagent intended for linker construction in PROTAC synthesis and bioconjugation.

  • Bifunctional PEG6 linker with two carboxylic acid termini.
  • High purity (≥98.0%) and consistent quality for synthesis.
  • Molecular weight 382.40 g/mol and formula C16H30O10.
  • Improves solubility and provides a flexible, hydrophilic spacer.
  • Supplied as a powder with defined storage recommendations for long-term stability.

Catalog No. 50-002-34386


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.