Learn More
Medchemexpress LLC Bis-PEG6-NHS ester | 1526718-98-8 | MFCD24539464 | 98.0% | 576.55 | C24H36N2O14 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1304105G
Encompass_Preferred
Bis-PEG6-NHS ester is a PEG6-based bifunctional NHS-ester linker used to assemble PROTACs and as a cleavable linker in antibody-drug conjugate synthesis. It reacts with primary amines to form amide bonds, and the PEG6 spacer improves hydrophilicity and flexibility.
  • Peg6 spacer provides flexibility and hydrophilicity.
  • NHS ester functional groups react with primary amines for conjugation.
  • Cleavable linker suitable for ADC and PROTAC synthesis.
  • High purity (98.0%) and molecular weight 576.55.
  • Soluble in DMSO at 125 mg/mL; ultrasonic recommended.
  • Hygroscopic; store sealed at -20°C and prepare solutions freshly.
Catalog No. 50-002-04126