Learn More
Strem, An Ascensus Company CAS# 11136-36-0. 1g. Bis(pentamethylcyclopentadienyl)titanium dichloride, 99%. MFCD00049153
SDP
Supplier:  Strem, An Ascensus Company 2203001G
Encompass
CAS# 11136-36-0. 1g. Bis(pentamethylcyclopentadienyl)titanium dichloride, 99%. MFCD00049153. Molecular Weight: 389.23. Molecular Formula: [(CH3)5C5]2TiCl2. Color/form: red-brown xtl. Strem# 22-0300. http://www.strem.com/catalog/v/22-0300/
Catalog No. NC1749451