Learn More
Medchemexpress LLC Bis-propargyl-PEG1 | 40842-04-4 | 98.0% | 138.16 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140038100MG
Encompass_Preferred
Bis-propargyl-PEG1 is a PEG-based PROTAC linker that facilitates the synthesis of PROTACs. This click chemistry reagent contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Suitable for PROTAC synthesis
Catalog No. 50-003-51375