Learn More
Medchemexpress LLC Bis-propargyl-PEG11 | 1351373-49-3 | 99.79% | 578.69 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140039
Encompass_Preferred
Bis-propargyl-PEG11 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups.
  • PEG-based PROTAC linker.
  • Can be used in the synthesis of PROTACs.
  • Click chemistry reagent with an Alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.
Catalog No. 50-003-51134