Learn More
Medchemexpress LLC Bis-propargyl-PEG4 | 159428-42-9 | 95.6% | 250 MG
SDP
Supplier:  Medchemexpress LLC HY120397250MG
Encompass_Preferred
Bis-propargyl-PEG4 is a PEG4-based bifunctional linker bearing terminal propargyl (alkyne) groups. It is designed for click chemistry and PROTAC linker synthesis, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) for conjugation to azide-bearing partners.
  • Peg4 linker with terminal propargyl (alkyne) groups.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation.
  • Suited for PROTAC synthesis and small-molecule dimer formation.
  • Defined molecular weight and formula for predictable stoichiometry.
  • Available in milligram to gram packaging for research use.
Catalog No. 50-002-34463