Learn More
Medchemexpress LLC Bis-propargyl-PEG6 1g | 400775-35-1 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1171861G
Encompass_Preferred
Bis-propargyl-PEG6 is a short PEG spacer with terminal propargyl (alkyne) groups used as a linker or crosslinker in click chemistry, bioconjugation, and PROTAC synthesis. It provides flexible spacing with reactive alkyne handles suitable for copper-catalyzed azide-alkyne cycloaddition and related conjugation reactions.
  • Terminal propargyl groups for click chemistry conjugation.
  • PEG6 spacer provides flexibility and water solubility.
  • Suitable for PROTAC linker design and bioconjugation workflows.
  • Offered in a 1 g research pack for small-scale synthesis.
Catalog No. 50-002-26357