Learn More
Medchemexpress LLC Bis-SS-C3-NHS ester | 98604-88-7 | 98.0% | 432.47 g/mol | C16H20N2O8S2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY133584250MG
Encompass_Preferred
Bis-SS-C3-NHS ester is a cleavable antibody-drug conjugate (ADC) linker containing N-hydroxysuccinimide (NHS) ester functional groups for amine coupling and a disulfide spacer that enables payload release under reductive intracellular conditions.
  • Cleavable disulfide spacer for intracellular payload release.
  • NHS ester functional groups for efficient amine coupling to biomolecules.
  • High purity suitable for conjugation chemistry.
  • White to off-white solid physical form.
  • Recommended storage at -20°C, sealed and protected from moisture; in solvent, -80°C for up to 6 months.
Catalog No. 50-002-03714