Learn More
Medchemexpress LLC Bis-SS-C3-NHS ester | 98604-88-7 | 98.0% | 432.47 g·mol⁻¹ | C16H20N2O8S2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1335845MG
Encompass_Preferred
Bis-SS-C3-NHS ester is a cleavable disulfide-containing NHS ester linker reagent used to conjugate amine-containing payloads during antibody-drug conjugate (ADC) synthesis. It provides a reducible spacer and an activated ester for efficient amide bond formation.
  • cleavable disulfide linker for controlled payload release
  • nhs ester functional group for amine conjugation
  • suitable for antibody-drug conjugate synthesis
  • reported high purity for reliable coupling reactions
  • store cold and protect from moisture to preserve stability
Catalog No. 50-002-03369