Learn More
Medchemexpress LLC Bis-Tos-PEG4 (PROTAC Linker 16) | 37860-51-8 | 502.60 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00481650G
Encompass_Preferred
Bis-Tos-PEG4 (PROTAC Linker 16) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets the protein of interest. These molecules leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • Utilizes intracellular ubiquitin-proteasome system
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-004-46026