Learn More
Medchemexpress LLC Bis-Tos-PEG6 | 42749-27-9 | C26H38O11S2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY140367500MG
Encompass_Preferred
Bis-Tos-PEG6 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) consist of two different ligands connected by a linker: one ligand for an E3 ubiquitin ligase and another for the target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs.
  • PEG-based PROTAC linker.
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-56597