Learn More
Medchemexpress LLC Bis-Tos-PEG7 | 69502-27-8 | C28H42O12S2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140368
Encompass_Preferred
Bis-Tos-PEG7 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs employ two distinct ligands joined by a linker: one targeting an E3 ubiquitin ligase and the other for a specific target protein. They harness the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-52581