Learn More
Medchemexpress LLC Bis-Tos-PEG7 | 69502-27-8 | C28H42O12S2 | 50 MG
Supplier: Medchemexpress LLC HY14036850MG
Bis-Tos-PEG7 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules with two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase, and the other targets a protein of interest. They exploit the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
- Peg-based protac linker
- Utilized in the synthesis of protacs
- Functions by leveraging the ubiquitin-proteasome system
- Selectively degrades specific target proteins