Learn More
Strem, An Ascensus Company CAS# 4808-30-4. 5g. Bis(tri-n-butyltin)sulfide, 97%. MFCD00048750
SDP
Supplier:  Strem, An Ascensus Company 9350025G
Encompass
CAS# 4808-30-4. 5g. Bis(tri-n-butyltin)sulfide, 97%. MFCD00048750. Molecular Weight: 612.14. Molecular Formula: [(C4H9)3Sn]2S. Color/form: colorless liq. Strem# 93-5002. http://www.strem.com/catalog/v/93-5002/
Catalog No. 50-901-15539