Learn More
Medchemexpress LLC Bis(trimethylsilyl)acetylene | 14630-40-1 | MFCD00008276 | 99.9% | 170.40 g/mol | C8H18Si2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01696150G
Encompass_Preferred
Bis(trimethylsilyl)acetylene is an organosilicon reagent used as a protected acetylene equivalent in organic synthesis and life-science research. It facilitates introduction of acetylene functionality while offering improved handling and stability compared with terminal acetylenes.
  • High purity suitable for synthetic applications (99.87%).
  • Protected acetylene equivalent for safer handling and improved stability.
  • Useful in coupling and cycloaddition reactions as an acetylene source.
  • Suitable as a building block for complex molecule synthesis.
  • Available in multiple pack sizes for lab-scale work.
Catalog No. 50-003-01932