Learn More
Strem, An Ascensus Company CAS# 13007-90-4. 5g. Bis(triphenylphosphine)nickel dicarbonyl, 98%. MFCD00009591
SDP
Supplier:  Strem, An Ascensus Company 2801005G
Encompass
CAS# 13007-90-4. 5g. Bis(triphenylphosphine)nickel dicarbonyl, 98%. MFCD00009591. Molecular Weight: 639.29. Molecular Formula: [(C6H5)3P]2Ni(CO)2. Color/form: light yellow xtl. Strem# 28-0100. http://www.strem.com/catalog/v/28-0100/
Catalog No. 50-901-11236