Learn More
Genscript Corporation SurePAGE™, Bis-Tris, 10x8, 10%, 10 wells
Supplier:  Genscript Corporation M00664
Encompass
GenScript SurePAGE, Bis-Tris, 10 cm x 8 cm gels are high-performance precast mini polyacrylamide gels with a special design that allows large sample loading volumes. The unique formulation of the gel and cassette design enables superior band resolution and significantly improved band evenness. SurePAGE gels are cast in a neutral pH buffer that minimizes polyacrylamide hydrolysis, increases gel stability and minimizes protein modification. SurePAGE gels guarantee excellent batch-to-batch consistency and a reliable protein migration pattern. With specially formulated Tris-MOPS running buffer, proteins can be separated quickly and efficiently for subsequent detection by staining or Western blotting. SurePAGE, Bis-Tris, 10x8 gels are available in gradient (4-20%, 4-12%, and 8-16%) and homogeneous (8%, 10%, 12% and 15%) concentrations. Each gel concentration has comb configurations of 10-well, 12-well and 15-well.
Catalog No. NC2192547