Learn More
Medchemexpress LLC Bis-Tris-d19 | 352534-93-1 | 99% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYD0856S
Encompass_Preferred
Bis-Tris-d19 is the deuterium labeled Bis-Tris, a commonly used buffer. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs. This compound is suitable for various analytical applications.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium-labeled compound
Catalog No. 50-004-11949