Learn More
Medchemexpress LLC Bis-Tris-d19 | 352534-93-1 | 99% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD0856S5MG
Encompass_Preferred
Bis-Tris-d19 is the deuterium labeled Bis-Tris, a commonly used buffer. It serves as a tracer or an internal standard for quantitative analysis through various methods including NMR, GC-MS, or LC-MS.
  • Deuterium labeled Bis-Tris
  • Functions as a tracer in analytical studies
  • Acts as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Solid, white to off-white appearance
  • Offers 99% purity
Catalog No. 50-004-10046