Learn More
Medchemexpress LLC Bis-Tris-d19 | 352534-93-1 | 99% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYD0856S50MG
Encompass_Preferred
Bis-Tris-d19 is the deuterium-labeled form of Bis-Tris, a commonly used buffer. Deuteration, which involves incorporating stable heavy isotopes into drug molecules, is used as tracers for quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09972