Learn More
Alkali Scientific BIS TRIS Propane [1,3 Bis (TRIS(hydroxymethyl)methylamino)propane], 500 G
SDP
Supplier:  Alkali Scientific IST005CP
Encompass_Preferred
BIS TRIS Propane (1,3 Bis (TRIS(hydroxymethyl)methylamino)propane) is a high-purity biological buffer commonly used in laboratory applications requiring precise pH control. This powder form of the buffer is highly soluble in water, allowing for easy preparation of buffered solutions with specific pH values. It is frequently used in protein electrophoresis, chromatography, and enzyme assays where stable pH is crucial for maintaining the activity and structure of biological molecules. The high purity of BIS TRIS Propane ensures minimal interference in sensitive biochemical analyses, making it ideal for use in research and clinical laboratories where accuracy is essential. The 500g size provides an ample supply for routine laboratory procedures.
Catalog No. 50-006-59752