Learn More
Sigma Aldrich Fine Chemicals Biosciences BIS-TRIS PROPANE,500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences B4679500G
Encompass
BIS-TRIS propane | 1,3-Bis[tris(hydroxymethyl)methylamino]propane | CAS No.: 64431-96-5 | Linear Formula:CH2[CH2NHC(CH2OH)3]2 | Molecular Weight: 282.33
Catalog No. NC2209968