Learn More
AdipoGen Bis4-iodophenyl-iodonium BF4
SDP
Supplier:  AdipoGen CDXI0053G001
Encompass_Preferred
Chemical. CAS 1260147-20-3. Formula C12H8I3 . BF4. MW 532.9 . 86.8. Synthetic. Diaryliodonium salts. Use for the synthesis of tri-, tetra-, and penta-substituted pyrrole compounds.
Catalog No. 50-256-0637