Learn More
AdipoGen Bis4methylphenyliodonium PF6
SDP
Supplier:  AdipoGen CDXB0247G010
Encompass_Preferred
Chemical. CAS 60565-88-0. Formula C14H14F6IP. MW 454.13. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-256-1625