Learn More
Medchemexpress LLC β-bisabolene | 495-61-4 | MFCD16876335 | 97.3% | C15H24 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13655210MG
Encompass_Preferred
β-Bisabolene is a naturally occurring sesquiterpene (C15H24, MW 204.35 g/mol) isolated from opoponax (Commiphora guidotti). It has reported anti-cancer activity and is used as a research reagent in preclinical and in vitro studies, including breast cancer models.
  • Natural sesquiterpene isolated from plant sources
  • Reported anti-cancer activity useful for oncology research
  • Suitable for in vitro and preclinical study applications
  • Molecular formula C15H24 and molecular weight 204.35 g/mol
  • High purity (approximately 97.3%) for research experiments
Catalog No. 50-002-59093