Learn More
Medchemexpress LLC Bisabolene (mixture of isomers) | 495-62-5 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1212625G
Encompass_Preferred
Bisabolene (mixture of isomers) is a sesquiterpene research chemical supplied as a mixture of structural isomers. It is used in biochemical studies, metabolic engineering research, and fragrance-related analyses. The product is provided in small pack sizes suitable for preparing concentrated stock solutions and analytical standards.
  • Sesquiterpene mixture with molecular formula C15H24 and molecular weight 204.35.
  • Suitable for biochemical and metabolic engineering studies.
  • Pack sizes intended for research-scale use and stock solution preparation.
  • Store under nitrogen at -20°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-32874