Learn More
Medchemexpress LLC Bisabolol oxide A | 22567-36-8 | 10 MG

Supplier: Medchemexpress LLC HYN811710MG
Bisabolol oxide A is a bisabolane-type sesquiterpenoid found in the flowers of Matricaria chamomilla. It contributes to the antispasmodic effect of chamomile preparations and serves as a structural reference to confirm the identity of 9-hydroxybisabolol oxide A as its hydroxylated derivative, using published NMR spectroscopic data.
- Bisabolane-type sesquiterpenoid
- Found in the flowers of Matricaria chamomilla
- Contributes to the antispasmodic effect of chamomile preparations
- Purity of 99.66%
- Molecular weight of 238.37
- Molecular formula: C15H26O2
- Appearance: Liquid
- Color: Colorless to light yellow
- Target: Biochemical assay reagents
- Pathway: Others
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.