Learn More
Medchemexpress LLC Bisabolol oxide A | 22567-36-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN81175MG
Encompass_Preferred
Bisabolol oxide A is a bisabolane-type sesquiterpenoid found in the flowers of *Matricaria chamomilla* (chamomile). It contributes to the antispasmodic effect of chamomile preparations and serves as a structural reference to confirm the identity of 9-hydroxybisabolol oxide A as its hydroxylated derivative.
  • Found in *Matricaria chamomilla* (chamomile) flowers
  • Contributes to antispasmodic effect of chamomile preparations
  • Serves as a structural reference
  • For research use only
  • Initial source: Plants Leguminosae *Distemonanthus benthamianus Baill.*, Plants Compositae
Catalog No. 50-004-17558