Learn More
Medchemexpress LLC Bisantrene dihydrochloride | 71439-68-4 | MFCD01743164 | 98.9% | 471.39 g/mol | C22H24Cl2N8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100875A10MG
Encompass_Preferred
Bisantrene dihydrochloride is the dihydrochloride salt of an anthracene-derived antitumor compound supplied as a solid for research use. It intercalates DNA and inhibits eukaryotic type II topoisomerases, making it useful in cytotoxicity, mechanism-of-action, and drug resistance studies.
Catalog No. 50-002-12525