Learn More
TARGETMOL CHEMICALS INC Bisaramil hydrochloride 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T268221MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bisaramil hydrochloride (Bisaramil) is a novel diazabicyclononane antiarrhythmic compound that inhibits the generation of free radicals.Bisaramil hydrochloride blocks sodium currents and inhibits the slow Ca(2+) action potential induced by isoproterenol in K(+)-depolarized muscle. Purity 99.64%
Catalog No. 50-287-9196