Learn More
Cayman Chemical BIsdemethoxycurcumIn 10mg
SDP
Supplier:  Cayman Chemical 1096010 MG
Encompass_Preferred
A natural demethoxy derivative of curcumin; activates macrophage phagocytosis and, with 1α,25-dihydroxy vitamin D3, stimulates amyloid-β clearance by macrophages (optimal stimulation at 100 nM BDMC); suppresses proliferation in cancer cells; down-regulates p300 and inhibits LPS-induced iNOS expression
Catalog No. 50-295-1210