Learn More
TARGETMOL CHEMICALS INC Bisindolylmaleimide I 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T651310MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bisindolylmaleimide I (GF109203X) is a potent and highly selective protein kinase C (PKC) inhibitor, exhibiting a Ki of 14 nM. Purity 99.08%
Catalog No. 50-287-6891