Learn More
Medchemexpress LLC Bisindolylmaleimide I | 133052-90-1 | 98.2% | 412.48 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1386750MG
Encompass_Preferred
Bisindolylmaleimide I is a cell-permeable and reversible protein kinase C (PKC) inhibitor, identified with IC50 values for various PKC isoforms. It also acts as a glycogen synthase kinase-3 (GSK-3) inhibitor and is for research use only.
  • Cell-permeable and reversible PKC inhibitor (IC50 for PKCα, PKCβI, PKCβII, PKCγ: 16-20 nM)
  • GSK-3 inhibitor
  • Inhibits human platelet aggregation
  • Protects against *Bacillus anthracis* lethal toxin-mediated cytotoxicity
  • Inhibits α-thrombin-induced P47 phosphorylation
  • Reduces GSK-3 activity in adipocyte lysates
  • Inhibits exosome and microvesicle release from PC3 cells
  • Enhances cytotoxicity of 5-fluorouracil
  • Demonstrates *in vivo* activity in mice and shrews
Catalog No. 50-003-50791