Learn More
Medchemexpress LLC Bisindolylmaleimide I | 133052-90-1 | MFCD00236428 | 98.2% | 412.48 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13867100MG
Encompass_Preferred
Bisindolylmaleimide I is a cell-permeable and reversible protein kinase C (PKC) inhibitor. It exhibits inhibitory activity against various PKC isoforms, including PKCα, PKCβI, PKCβII, and PKCγ. This compound also functions as a GSK-3 inhibitor, making it suitable for a range of research applications.
  • Cell-permeable and reversible PKC inhibitor
  • Inhibits PKCα with an IC50 of 20 nM
  • Inhibits PKCβI with an IC50 of 17 nM
  • Inhibits PKCβII with an IC50 of 16 nM
  • Inhibits PKCγ with an IC50 of 20 nM
  • Functions as a GSK-3 inhibitor
  • Protects against *Bacillus anthracis* lethal toxin-mediated cytotoxicity
  • Inhibits exosome and microvesicle release from PC3 cells
  • Enhances cytotoxicity of 5-fluorouracil
Catalog No. 50-003-49883