Learn More
Medchemexpress LLC Bisindolylmaleimide I | 133052-90-1 | MFCD00236428 | 98.2% | C25H24N4O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY1386710MG
Encompass_Preferred
Bisindolylmaleimide I is a small-molecule, cell-permeable, reversible inhibitor of protein kinase C (PKC) used as a biochemical tool in cell-free and cell-based research to probe PKC-dependent signaling pathways. It is supplied as a solid powder and commonly stored at -20 °C for long-term stability.
  • Potent inhibitor of protein kinase C with low-nanomolar IC50 values for common isoforms.
  • Cell-permeable and reversible, suitable for cellular assays.
  • Useful in both cell-free biochemical assays and cell-based studies.
  • Supplied as a stable solid, compatible with standard laboratory storage.
  • Available in small research pack sizes for analytical and experimental use.
Catalog No. 50-002-61611