Learn More
Selleck Chemical LLC Bisindolylmaleimide I (GF109203X) 1mg 133052-90-1 GO 6850
SDP
Supplier:  Selleck Chemical LLC S72081MG
Encompass_Preferred
Bisindolylmaleimide I (GF109203X, GO 6850) is a potent PKC inhibitor with IC50 of 20 nM, 17 nM, 16 nM, and 20 nM for PKC?, PKC?I, PKC?II, and PKC? in cell-free assays, respectively, showing more than 3000-fold selectivity for PKC as compared to EGFR, PDGFR and insulin receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6338