Learn More
Cayman Chemical BIsIndolylmaleImIde I 50mg
SDP
Supplier:  Cayman Chemical 1329850 MG
Encompass_Preferred
A highly selective, cell-permeable, and reversible PKC inhibitor (Ki = 14 nM); competitive inhibitor for the ATP binding site of PKC; highly selective for PKCα, β1, β2, γ, δ, and ε isozymes; inhibits GSK3 in primary adipocyte lysates (IC50 = 360 nM) and in GSK3β immunoprecipitates derived from rat epididymal adipocytes (IC50 = 170 nM); competitively antagonizes the 5-HT3 receptor with a Ki value of 61 nM
Catalog No. 50-295-2922