Learn More
Medchemexpress LLC Bisindolylmaleimide I hydrochloride | 176504-36-2 | 98.4% | 448.94 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13867A100MG
Encompass_Preferred
Bisindolylmaleimide I hydrochloride is a cell-permeable and reversible protein kinase C (PKC) inhibitor with IC50 values of 20 nM, 17 nM, 16 nM, and 20 nM for PKCα, PKCβI, PKCβII, and PKCγ, respectively. It also acts as a glycogen synthase kinase-3 (GSK-3) inhibitor, making it suitable for a wide range of research applications.
  • Cell-permeable and reversible protein kinase C inhibitor
  • Glycogen synthase kinase-3 inhibitor
  • Inhibits α-thrombin-induced P47 phosphorylation
  • Inhibits DNA synthesis in quiescent swiss 3T3 cells
  • Reduces GSK-3 activity in adipocyte lysates
  • Inhibits exosome and microvesicle release from PC3 cells
  • Enhances cytotoxicity of 5-fluorouracil
  • Reduces increased NLRP3, P-PKCɑ, and PKCɑ levels in mechanical ventilation group
  • Reduces Quinpirole-induced vomiting in shrews
  • Blocks Quinpirole-mediated ERK1/2 phosphorylation in shrew brainstems
Catalog No. 50-003-49864