Learn More
Medchemexpress LLC Bisindolylmaleimide I hydrochloride | 176504-36-2 | 98.4% | C25H25ClN4O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13867A10MG
Encompass_Preferred
Bisindolylmaleimide I hydrochloride is a research-grade protein kinase C (PKC) inhibitor supplied as the hydrochloride salt for laboratory use. It is intended for biochemical and cell-based assays and for analytical applications; not for human or veterinary use.
  • Reversible, cell-permeable inhibitor of PKC with nanomolar potency.
  • Suitable for biochemical and cell-based assays.
  • Hydrochloride salt improves solubility in aqueous media.
  • High reported purity for analytical applications.
  • Intended for research use only and method development.
Catalog No. 50-002-61625